(S)-N-(1-(N-Cyano-N'-(3-(difluoromethoxy)phenyl)carbamimidoyl)-3-(3,4-dichlorophenyl)-4,5-dihydro-1H-pyrazol-4-yl)-N-ethyl-2-hydroxyacetamide ≥99%

Supplier: Ambeed
Warning

Synonyms: BAY-598

A580882-1MG A580882-5MG A580882-100MG
76649-940EA 41.38 USD
76649-940 76649-942 77572-852
(S)-N-(1-(N-Cyano-N'-(3-(difluoromethoxy)phenyl)carbamimidoyl)-3-(3,4-dichlorophenyl)-4,5-dihydro-1H-pyrazol-4-yl)-N-ethyl-2-hydroxyacetamide ≥99%
(S)-N-(1-(N-Cyano-N'-(3-(difluoromethoxy)phenyl)carbamimidoyl)-3-(3,4-dichlorophenyl)-4,5-dihydro-1H-pyrazol-4-yl)-N-ethyl-2-hydroxyacetamide

Precaución: for Research Use Only

Formula: C₂₂H₂₀Cl₂F₂N₆O₃
MW: 525.34 g/mol
MDL Number: MFCD30146419
CAS Number: 1906919-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR