(R)-7-(3,4-Difluorobenzyl)-6-(methoxymethyl)-2-(5-methyl-2-((1-methyl-1H-pyrazol-5-yl)amino)pyrimidin-4-yl)-6,7-dihydroimidazo[1,2-a]pyrazin-8(5H)-one 98+%
Supplier: Ambeed
|
Warning
|
A580222-1MG
A580222-5MG
76649-936EA
62
USD
76649-936
76649-938
(R)-7-(3,4-Difluorobenzyl)-6-(methoxymethyl)-2-(5-methyl-2-((1-methyl-1H-pyrazol-5-yl)amino)pyrimidin-4-yl)-6,7-dihydroimidazo[1,2-a]pyrazin-8(5H)-one 98+%
(R)-7-(3,4-Difluorobenzyl)-6-(methoxymethyl)-2-(5-methyl-2-((1-methyl-1H-pyrazol-5-yl)amino)pyrimidin-4-yl)-6,7-dihydroimidazo[1,2-a]pyrazin-8(5H)-one
Precaución: for Research Use Only
Formula:
C₂₄H₂₄F₂N₈O₂ |
MDL Number:
MFCD31631580 CAS Number: 2097416-76-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...