(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol hydrochloride 98%

Supplier: Ambeed
Warning

A578640-100MG A578640-1G A578640-250MG
76649-932EA 115.64 USD
76649-932 76649-934 77622-376
(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol hydrochloride 98%
(3-Aminobicyclo[1.1.1]pentan-1-yl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO·ClH
MW: 149.619 g/mol
MDL Number: MFCD27996088
CAS Number: 1638767-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR