4-Chloro-2-formylbenzoic acid 95%

Supplier: Ambeed
Warning

A577377-100MG A577377-250MG A577377-1G
76649-922EA 105.03 USD
76649-922 76649-924 77749-186
4-Chloro-2-formylbenzoic acid 95%
4-Chloro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₃
MW: 184.5765 g/mol
MDL Number: MFCD18453660
CAS Number: 4657-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR