3,5-Diamino-2-hydroxybenzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A576715-1G A576715-250MG A576715-5G
76649-914EA 117.76 USD
76649-914 76649-916 76649-918
3,5-Diamino-2-hydroxybenzoic acid hydrochloride 97%
3,5-Diamino-2-hydroxybenzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O₃
MW: 204.6100 g/mol
CAS Number: 177960-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR