[1,2,4]Triazolo[1,5-a]pyridin-6-ol 95%

Supplier: Ambeed
Warning

A574756-250MG A574756-50MG A574756-100MG A574756-1G
76649-906EA 132 USD
76649-906 76649-908 77617-430 77749-181
[1,2,4]Triazolo[1,5-a]pyridin-6-ol 95%
[1,2,4]Triazolo[1,5-a]pyridin-6-ol

Precaución: for Research Use Only

Formula: C₆H₅N₃O
MDL Number: MFCD22394729
CAS Number: 1394969-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR