Methyl 4-fluoro-2-formylbenzoate 95%

Supplier: Ambeed
Danger

A572935-250MG A572935-1G A572935-100MG
76649-892EA 157.02 USD
76649-892 77569-098 77617-420
Methyl 4-fluoro-2-formylbenzoate 95%
Methyl 4-fluoro-2-formylbenzoate

Precaución: For research use only.

Formula: C₉H₇FO₃
MW: 182.1485 g/mol
MDL Number: MFCD24685411
CAS Number: 106014-25-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR