Hydroxy-PEG1-CH2CO2tBu 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(2-hydroxyethoxy)acetate, 2-Methyl-2-propanyl (2-hydroxyethoxy)acetate, Hydroxy-PEG1-CH2-Boc

A572479-1G A572479-250MG A572479-5G A572479-100MG
76649-886EA 81.69 USD
76649-886 76649-888 76649-890 77617-418
Hydroxy-PEG1-CH2CO2tBu 98%
Hydroxy-PEG1-CH2CO2tBu

Precaución: for Research Use Only

Formula: C₈H₁₆O₄
MW: 176.21 g/mol
MDL Number: MFCD27635155
CAS Number: 287174-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR