(S)-tert-Butyl (1-(methoxy(methyl)amino)-1-oxopropan-2-yl)(methyl)carbamate 97%

Supplier: Ambeed
Warning

A570585-100MG A570585-250MG
76649-870EA 6 USD
76649-870 76649-872
(S)-tert-Butyl (1-(methoxy(methyl)amino)-1-oxopropan-2-yl)(methyl)carbamate 97%
(S)-tert-Butyl (1-(methoxy(methyl)amino)-1-oxopropan-2-yl)(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₄
MW: 246.3034 g/mol
MDL Number: MFCD31729566
CAS Number: 170097-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR