3-(3,5-Bis(trifluoromethyl)phenyl)-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A570273-5G A570273-1G
76649-868EA 112.46 USD
76649-868 77551-720
3-(3,5-Bis(trifluoromethyl)phenyl)-1H-1,2,4-triazole 98%
3-(3,5-Bis(trifluoromethyl)phenyl)-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₁₀H₅F₆N₃
MW: 281.1572 g/mol
MDL Number: MFCD31540669
CAS Number: 1333154-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR