4-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine 98%

Supplier: Ambeed
Warning

A565543-1G A565543-250MG A565543-5G
76649-854EA 78.51 USD
76649-854 76649-856 76649-858
4-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine 98%
4-(2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₇H₂₅BClNO₃
MW: 337.6493 g/mol
MDL Number: MFCD20441908
CAS Number: 2096337-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR