5-(Di(adamantan-1-yl)phosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole 97%
Supplier: Ambeed
|
Warning
|
A564640-100MG
A564640-1G
A564640-250MG
A564640-5G
76649-846EA
92
USD
76649-846
76649-848
76649-850
76649-852
5-(Di(adamantan-1-yl)phosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole 97%
5-(Di(adamantan-1-yl)phosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole
Precaución: for Research Use Only
Formula:
C₄₄H₄₇N₄P MW: 662.8446 g/mol |
MDL Number:
MFCD22200502 CAS Number: 1239478-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...