4-Oxo-6-((pyrimidin-2-ylthio)methyl)-4H-pyran-3-yl 4-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: ML221

A564295-10MG A564295-1G A564295-1MG A564295-25MG A564295-5MG A564295-100MG A564295-250MG A564295-5G
76649-836EA 18 USD
76649-836 76649-838 76649-840 76649-842 76649-844 77617-394 77617-396 77617-398
4-Oxo-6-((pyrimidin-2-ylthio)methyl)-4H-pyran-3-yl 4-nitrobenzoate 97%
4-Oxo-6-((pyrimidin-2-ylthio)methyl)-4H-pyran-3-yl 4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₁N₃O₆S
MDL Number: MFCD06407006
CAS Number: 877636-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR