(Aminomethyl)ferrocene hydrochloride 98%

Supplier: Ambeed
Warning

A563427-100MG A563427-1G A563427-250MG
76649-826EA 33.95 USD
76649-826 76649-828 76649-830
(Aminomethyl)ferrocene hydrochloride 98%
(Aminomethyl)ferrocene hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₅ClFeN
MW: 242.4621 g/mol
MDL Number: MFCD32011687
CAS Number: 12261-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR