Ethyl 2-bromo-5-fluoroisonicotinate 95%

Supplier: Ambeed
Warning

A562468-1G A562468-250MG
76649-816EA 23 USD
76649-816 76649-818
Ethyl 2-bromo-5-fluoroisonicotinate 95%
Ethyl 2-bromo-5-fluoroisonicotinate

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
MDL Number: MFCD14698321
CAS Number: 1072206-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR