3-Chloro-6,7-dihydro-5H-cyclopenta[c]pyridine-4-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A561296-100MG
A561296-1G
A561296-250MG
A561296-5G
76649-800EA
158.08
USD
76649-800
76649-802
76649-804
77749-158
3-Chloro-6,7-dihydro-5H-cyclopenta[c]pyridine-4-carbonitrile 95%
3-Chloro-6,7-dihydro-5H-cyclopenta[c]pyridine-4-carbonitrile
Precaución: for Research Use Only
Formula:
C₉H₇ClN₂ MW: 178.6183 g/mol |
MDL Number:
MFCD19690416 CAS Number: 103589-67-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...