Methyl 2-amino-6-chloroisonicotinate 97%

Supplier: Ambeed
Danger

A560920-1G A560920-5G
76649-788EA 222.79 USD
76649-788 77598-570
Methyl 2-amino-6-chloroisonicotinate 97%
Methyl 2-amino-6-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.5957 g/mol
MDL Number: MFCD18447558
CAS Number: 1005508-80-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR