14-((2,5-Dioxopyrrolidin-1-yl)oxy)-14-oxotetradecanoic acid 95%

Supplier: Ambeed
Warning

A560830-100MG A560830-1G A560830-250MG
76649-782EA 94.43 USD
76649-782 76649-784 76649-786
14-((2,5-Dioxopyrrolidin-1-yl)oxy)-14-oxotetradecanoic acid 95%
14-((2,5-Dioxopyrrolidin-1-yl)oxy)-14-oxotetradecanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₉NO₆
MW: 355.4260 g/mol
CAS Number: 146004-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR