Benzyl (S)-2-(cyanomethyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A559651-100MG A559651-250MG A559651-1G
76649-778EA 27.59 USD
76649-778 76649-780 77572-768
Benzyl (S)-2-(cyanomethyl)piperazine-1-carboxylate 97%
Benzyl (S)-2-(cyanomethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₇N₃O₂
MW: 259.3037 g/mol
MDL Number: MFCD31728479
CAS Number: 2158302-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR