1-(Methylsulfonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline 95%

Supplier: Ambeed
Warning

A558806-100MG A558806-1G
76649-774EA 39.26 USD
76649-774 76649-776
1-(Methylsulfonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline 95%
1-(Methylsulfonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₄S
MW: 323.2155 g/mol
MDL Number: MFCD28718122
CAS Number: 1132944-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR