5-Chloro-2,4-difluoropyridine 95%

Supplier: Ambeed
Danger

A558794-1G A558794-250MG
76649-770EA 1131.99 USD
76649-770 76649-772
5-Chloro-2,4-difluoropyridine 95%
5-Chloro-2,4-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MW: 149.5259 g/mol
MDL Number: MFCD22577173
CAS Number: 1807257-52-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR