4-Methyl-6-nitropyridin-3-ol 98%

Supplier: Ambeed
Warning

A556533-100MG A556533-250MG
76649-764EA 28 USD
76649-764 76649-766
4-Methyl-6-nitropyridin-3-ol 98%
4-Methyl-6-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MDL Number: MFCD28799553
CAS Number: 1806389-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR