1,2-Di-p-tolylethyne 97%

Supplier: Ambeed
Warning

A556416-100MG A556416-1G A556416-250MG
76649-760EA 31.83 USD
76649-760 76649-762 77622-362
1,2-Di-p-tolylethyne 97%
1,2-Di-p-tolylethyne

Precaución: for Research Use Only

Formula: C₁₆H₁₄
MW: 206.2824 g/mol
MDL Number: MFCD00010651
CAS Number: 2789-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR