ω-Bromo-2,4-dimethylacetophenone 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(2,4-dimethylphenyl)ethanone, α-Bromo-2,4-dimethylacetophenone, 2-Bromo-2',4'-dimethylacetophenone

A554973-250MG A554973-5G
76649-752EA 22.28 USD
76649-752 76649-754
ω-Bromo-2,4-dimethylacetophenone 98%
ω-Bromo-2,4-dimethylacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
MW: 227.1 g/mol
MDL Number: MFCD00017872
CAS Number: 26346-85-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR