3-Phenyl-1H-pyrazole-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A554173-1G
A554173-250MG
A554173-5G
76649-746EA
26.53
USD
76649-746
76649-748
76649-750
3-Phenyl-1H-pyrazole-5-carboxylic acid 95%
3-Phenyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O₂ MW: 188.1800 g/mol |
MDL Number:
MFCD01248821 CAS Number: 1134-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...