6-Fluorobenzofuran-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A554012-100MG A554012-1G A554012-250MG
76649-740EA 42.44 USD
76649-740 76649-742 76649-744
6-Fluorobenzofuran-3-carboxylic acid 98%
6-Fluorobenzofuran-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅FO₃
MW: 180.133 g/mol
MDL Number: MFCD22544066
CAS Number: 1393561-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR