Methyl 2-bromo-4-fluoro-5-nitrobenzoate 98%

Supplier: Ambeed
Danger

A551530-100MG A551530-1G A551530-250MG
76649-728EA 47.75 USD
76649-728 76649-730 76649-732
Methyl 2-bromo-4-fluoro-5-nitrobenzoate 98%
Methyl 2-bromo-4-fluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrFNO₄
MW: 278.0320 g/mol
MDL Number: MFCD12172996
CAS Number: 85953-31-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR