N-(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)cyclohexanamine 96%

Supplier: Ambeed
Danger

A551095-100MG A551095-1G A551095-250MG A551095-5G
76649-720EA 35.01 USD
76649-720 76649-722 76649-724 76649-726
N-(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)cyclohexanamine 96%
N-(3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)cyclohexanamine

Precaución: for Research Use Only

Formula: C₁₉H₂₉BClNO₂
MW: 349.7031 g/mol
MDL Number: MFCD18311895
CAS Number: 2096334-42-6
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR