1-(3-Bromo-6-(trifluoromethyl)pyridin-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A548150-1G A548150-250MG
76649-710EA 195 USD
76649-710 76649-712
1-(3-Bromo-6-(trifluoromethyl)pyridin-2-yl)ethanone 97%
1-(3-Bromo-6-(trifluoromethyl)pyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrF₃NO
MW: 268.0306 g/mol
CAS Number: 2384823-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR