2-Bromo-N-methylpyrimidin-4-amine 95%

Supplier: Ambeed
Danger

A548092-100MG A548092-1G A548092-250MG A548092-5G
76649-702EA 17 USD
76649-702 76649-704 76649-706 76649-708
2-Bromo-N-methylpyrimidin-4-amine 95%
2-Bromo-N-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MDL Number: MFCD14702740
CAS Number: 1209458-26-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR