NBQX 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo(f)-quinoxaline, 6-nitro-7-sulphamoylbenzo(f)-quinoxaline-2,3-dione, 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione, NBQX, 6-Nitro-2,3-dioxo-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulphonamide

A546099-10MG A546099-1MG A546099-25MG A546099-50MG A546099-5MG
76649-692EA 66.84 USD
76649-692 76649-694 76649-696 76649-698 76649-700
NBQX 97%
NBQX

Precaución: for Research Use Only

Formula: C₁₂H₈N₄O₆S
MW: 336.28 g/mol
MDL Number: MFCD00270054
CAS Number: 118876-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR