(E)-Cyclooct-4-en-1-yl (2,5-dioxopyrrolidin-1-yl) carbonate 95% rel-(1R-4E-pR)-equatorial

Supplier: Ambeed
Warning

Synonyms: TCO-NHS ester

A545750-25MG A545750-100MG A545750-10MG A545750-250MG A545750-1G A545750-50MG
76649-690EA 316.15 USD
76649-690 77598-556 77598-558 77598-560 77617-326 77617-328
(E)-Cyclooct-4-en-1-yl (2,5-dioxopyrrolidin-1-yl) carbonate 95% rel-(1R-4E-pR)-equatorial
(E)-Cyclooct-4-en-1-yl (2,5-dioxopyrrolidin-1-yl) carbonate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₅
MW: 267.2778 g/mol
MDL Number: MFCD24386372
CAS Number: 1191901-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR