tert-Butyl ((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Val-Cit-PAB
A542750-10MG
A542750-1MG
A542750-25MG
A542750-50MG
A542750-1G
A542750-5G
A542750-250MG
A542750-100MG
76649-678EA
28
USD
76649-678
76649-680
76649-682
76649-684
77569-058
77572-722
77598-554
77749-132
tert-Butyl ((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)carbamate 97%
tert-Butyl ((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)carbamate
Precaución: For research use only.
Formula:
C₂₃H₃₇N₅O₆ |
MDL Number:
MFCD22200277 CAS Number: 870487-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...