1-(2-Amino-5-bromopyridin-3-yl)ethanone 98%

Supplier: Ambeed
Warning

A542388-100MG A542388-1G A542388-250MG A542388-5G
76649-664EA 47 USD
76649-664 76649-666 76649-668 76649-670
1-(2-Amino-5-bromopyridin-3-yl)ethanone 98%
1-(2-Amino-5-bromopyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MDL Number: MFCD13190081
CAS Number: 1196154-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR