1-(5-Bromo-4-chloro-2-nitrophenyl)ethanone 95%
Supplier: Ambeed
|
Warning
|
A541631-100MG
A541631-1G
A541631-250MG
76649-650EA
135.8
USD
76649-650
76649-652
76649-654
1-(5-Bromo-4-chloro-2-nitrophenyl)ethanone 95%
1-(5-Bromo-4-chloro-2-nitrophenyl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₅BrClNO₃ MW: 278.4872 g/mol |
MDL Number:
MFCD30188735 CAS Number: 2092803-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...