2-Amino-3-((tert-butyldiphenylsilyl)oxy)-2-methylpropan-1-ol 95%

Supplier: Ambeed
Warning

A541452-1G A541452-250MG A541452-5G
76649-644EA 89 USD
76649-644 76649-646 76649-648
2-Amino-3-((tert-butyldiphenylsilyl)oxy)-2-methylpropan-1-ol 95%
2-Amino-3-((tert-butyldiphenylsilyl)oxy)-2-methylpropan-1-ol

Precaución: for Research Use Only

Formula: C₂₀H₂₉NO₂Si
MDL Number: MFCD30527952
CAS Number: 280753-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR