4-((2-(Dimethylamino)ethoxy)(phenyl)methyl)aniline 95%

Supplier: Ambeed
Warning

A540906-1G A540906-250MG
76649-640EA 493.32 USD
76649-640 76649-642
4-((2-(Dimethylamino)ethoxy)(phenyl)methyl)aniline 95%
4-((2-(Dimethylamino)ethoxy)(phenyl)methyl)aniline

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O
MW: 270.3694 g/mol
MDL Number: MFCD22373419
CAS Number: 101602-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR