5-Ethynyl-2-fluoroaniline 95%

Supplier: Ambeed
Warning

A540270-100MG A540270-250MG A540270-1G
76649-636EA 29 USD
76649-636 76649-638 77749-129
5-Ethynyl-2-fluoroaniline 95%
5-Ethynyl-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.1400 g/mol
MDL Number: MFCD11036139
CAS Number: 1010422-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR