Di-n-octyl sebacate 98%

Supplier: Ambeed
Warning

Synonyms: Sebacic Acid Di-n-octyl Ester, Decanedioic Acid Di-n-octyl Ester, Di-n-octyl Decanedioate

A539810-100G A539810-500G
76649-632EA 29.71 USD
76649-632 76649-634
Di-n-octyl sebacate 98%
Di-n-octyl sebacate

Precaución: for Research Use Only

Formula: C₂₆H₅₀O₄
MW: 426.68 g/mol
MDL Number: MFCD00059269
CAS Number: 14491-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR