3-Chloroimidazo[1,5-a]pyridine 95%

Supplier: Ambeed
Danger

A538919-100MG A538919-250MG A538919-50MG A538919-1G
76649-626EA 165.51 USD
76649-626 76649-628 76649-630 77598-552
3-Chloroimidazo[1,5-a]pyridine 95%
3-Chloroimidazo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MDL Number: MFCD30067670
CAS Number: 1895224-02-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR