N-Methyl-3-(trifluoromethyl)cyclobutan-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A537571-100MG A537571-250MG
76649-622EA 294.94 USD
76649-622 76649-624
N-Methyl-3-(trifluoromethyl)cyclobutan-1-amine hydrochloride 98%
N-Methyl-3-(trifluoromethyl)cyclobutan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClF₃N
MW: 189.6065 g/mol
MDL Number: MFCD31705847
CAS Number: 2231675-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR