CEP-40783 97%

Supplier: Ambeed
Warning

Synonyms: N-(4-((6,7-Dimethoxyquinolin-4-yl)oxy)-3-fluorophenyl)-3-(4-fluorophenyl)-1-isopropyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxamide

A525104-1MG A525104-2MG A525104-50MG A525104-5MG
76649-584EA 12.74 USD
76649-584 76649-586 76649-588 76649-590
CEP-40783 97%
CEP-40783

Precaución: for Research Use Only

Formula: C₃₁H₂₆F₂N₄O₆
MW: 588.56 g/mol
MDL Number: MFCD28502441
CAS Number: 1437321-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR