5-(4-Methoxyphenyl)-3-methyl-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A523007-100MG A523007-250MG
76649-576EA 59.42 USD
76649-576 76649-578
5-(4-Methoxyphenyl)-3-methyl-1H-pyrrole-2-carboxylic acid 97%
5-(4-Methoxyphenyl)-3-methyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₃
MW: 231.2500 g/mol
CAS Number: 2106310-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR