3-Bromoimidazo[1,2-a]pyridin-8-ol 95%

Supplier: Ambeed
Warning

A520583-250MG A520583-5G
76649-566EA 24.41 USD
76649-566 76649-568
3-Bromoimidazo[1,2-a]pyridin-8-ol 95%
3-Bromoimidazo[1,2-a]pyridin-8-ol

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MW: 213.03 g/mol
MDL Number: MFCD31698829
CAS Number: 1780873-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR