6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiazolo[5,4-b]pyridine 97%

Supplier: Ambeed
Warning

A516315-100MG A516315-250MG A516315-50MG A516315-1G
76649-552EA 152.77 USD
76649-552 76649-554 76649-556 77749-069
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiazolo[5,4-b]pyridine 97%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiazolo[5,4-b]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₂S
MW: 262.14 g/mol
MDL Number: MFCD22544252
CAS Number: 1580489-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR