ND-630 97%

Supplier: Ambeed
Warning

Synonyms: Firsocostat, GS0976, (R)-2-(1-(2-(2-Methoxyphenyl)-2-((tetrahydro-2H-pyran-4-yl)oxy)ethyl)-5-methyl-6-(oxazol-2-yl)-2,4-dioxo-1,4-dihydrothieno[2,3-d]pyrimidin-3(2H)-yl)-2-methylpropanoic acid, 1,4-dihydro-1-[(2R)-2-(2-methoxyphenyl)-2-[(tetrahydro-2H-pyran-4-yl)oxy]ethyl]-α,α,5-trimethyl-6-(2-oxazolyl)-2,4-dioxo-thieno[2,3-d]pyrimidine-3(2H)-acetic acid

A512642-50MG A512642-5MG A512642-10MG A512642-1G A512642-100MG A512642-250MG
76649-520EA 168.69 USD
76649-520 76649-522 77749-054 77749-055 77749-053 77749-056
ND-630 97%
ND-630

Precaución: for Research Use Only

Formula: C₂₈H₃₁N₃O₈S
MW: 569.63 g/mol
MDL Number: MFCD28963986
CAS Number: 1434635-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR