TCE35031 95%

Supplier: Ambeed
Warning

Synonyms: 2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetic acid, Thalidomide-O-COOH

A511927-1MG A511927-250MG A511927-5G A511927-1G A511927-100MG A511927-25G
76649-510EA 10.61 USD
76649-510 76649-512 76649-514 77551-536 77598-536 77617-186
TCE35031 95%
TCE35031

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₇
MW: 332.26 g/mol
MDL Number: MFCD31560477
CAS Number: 1061605-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR