2-(4-(1,2,2-triphenylvinyl)phenoxy)acetic acid 98% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A510968-100MG
A510968-1G
A510968-250MG
A510968-5G
76649-502EA
54.11
USD
76649-502
76649-504
76649-506
76649-508
2-(4-(1,2,2-triphenylvinyl)phenoxy)acetic acid 98% stabilized
2-(4-(1,2,2-triphenylvinyl)phenoxy)acetic acid
Precaución: for Research Use Only
Formula:
C₂₈H₂₂O₃ MW: 406.4725 g/mol |
MDL Number:
MFCD30721610 CAS Number: 1471339-65-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...