2-Bromo-6-(methoxymethoxy)naphthalene 97%

Supplier: Ambeed
Warning

A510133-10G A510133-1G A510133-250MG A510133-5G
76649-494EA 63 USD
76649-494 76649-496 76649-498 76649-500
2-Bromo-6-(methoxymethoxy)naphthalene 97%
2-Bromo-6-(methoxymethoxy)naphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrO₂
CAS Number: 111359-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR