8-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A509603-100MG A509603-250MG
76649-484EA 352.22 USD
76649-484 77594-600
8-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 95%
8-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.0965 g/mol
MDL Number: MFCD21879585
CAS Number: 1368622-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR